3-Thiophencarbonitril
Lieferant: Thermo Scientific
Synonyme:
3-Thienylcyanid, Thiophen-3-carbonitril, Thiophen-3-carbonsäurenitril, 3-Cyanthiophen, 3-Thiophencarbonsäurenitril
SEW03680.250
SEW03680.1
MAYBSEW03680.250EA
0
EUR
MAYBSEW03680.250
MAYBSEW03680.1
3-Thiophencarbonitril
3-Thiophencarbonitril
Formel:
C₅H₃NS Molecular Weight: 109.15 g/mol Siedepunkt: 82 Dichte: 1.1956 g/cm³ (20 °C) |
MDL:
MFCD00151852 CAS-Nummer: 1641-09-4 EINECS: 216-687-7 UN: 3276 ADR: 6.1 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...