Eriochromblauschwarz R (Calcon®, Mordant Black 17), rein
Lieferant: Thermo Scientific
Achtung
Synonyme: 1-(2-Hydroxy-1-naphthylazo)- 2-naphthol-4-sulfonsäure Natriumsalz, Natrium-3-hydroxy-4-(2-hydroxy-1-naphthylazo)-1-naphthalinsulfonat, Eriochromblauschwarz R, Palatinchromschwarz 6 BN, Calcon®
202440250 202441000
ACRO202440250EA 31.2 EUR
ACRO202440250 ACRO202441000
Eriochromblauschwarz R (Calcon®, Mordant Black 17), rein
Eriochromblauschwarz R (Calcon®, Mordant Black 17)
Formel: C₂₀H₁₃N₂NaO₅S
Molecular Weight: 416,38 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00004069
CAS-Nummer: 2538-85-4
EINECS: 219-810-2
Bestellen
Spezifikation Testergebnisse
Appearance Dark brown to black Powder
Infrared spectrum Conforms
UV spectrum Complies