3-Thiophenboronsäure 97%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
3-Thienylboronsäure, Thiophen-3-boronsäure
345660010
345660050
ACRO345660010EA
31.5
EUR
ACRO345660010
ACRO345660050
3-Thiophenboronsäure 97%
3-Thiopheneboronic acid
Formel:
C₄H₅BO₂S Molecular Weight: 127,96 g/mol Schmelzpunkt: 162…167 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD00151851 CAS-Nummer: 6165-69-1 |
Spezifikation Testergebnisse
Appearance | White to beige to grey Powder |
Infrared spectrum | Conforms |
HPLC | ≥96.0 % |
Titration with NaOH | 96.0 to 106.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...