3-Thiophenboronsäure ≥98%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
3-Thienylboronsäure, Thiophen-3-boronsäure
B23637.06
B23637.03
B23637.14
B23637.06EA
93.4
EUR
B23637.06
B23637.03
B23637.14
3-Thiophenboronsäure ≥98%
3-Thiopheneboronic acid
Purity: 98%
CAS No.: 6165-69-1
Molecular Formula: C4H5BO2S
Formula Weight: 127.96
Storage Temperature: -30°C to -10°C
Melting Point: 164-169°
Beilstein Reference: 113573
MDL No.: MFCD00151851
Formel:
C₄H₅BO₂S Molecular Weight: 127,96 g/mol Schmelzpunkt: 164…169 °C |
MDL:
MFCD00151851 CAS-Nummer: 6165-69-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...