Bromkresolpurpur 0,4 g/l in Ethanol
Lieferant: Bernd Kraft
|
Gefahr
|
Synonyme:
5′,5''-Dibrom-o-kresolsulfonphthalein
03092.0000
03092.0700
03092.1700
KRAF03092.0000EA
0
EUR
KRAF03092.0000
KRAF03092.0700
KRAF03092.1700
Bromkresolpurpur 0,4 g/l in Ethanol
Bromkresolpurpur
Formel:
C₂₁H₁₆Br₂O₅S Molecular Weight: 540,23 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00011681 CAS-Nummer: 115-40-2 EINECS: 204-087-8 UN: 1170 ADR: 3,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...