5-Chlor-2-thiophenboronsäure
Lieferant: FLUOROCHEM
Synonyme:
5-Chlor-2-thienylboronsäure, 5-Chlorthiophen-2-boronsäure
010987-5G
010987-25G
FLUO010987-5GEA
115
EUR
FLUO010987-5G
FLUO010987-25G
5-Chlor-2-thiophenboronsäure
5-Chloro-2-thiopheneboronic acid
Formel:
C₄H₄BClO₂S Molecular Weight: 162,4 g/mol |
MDL:
MFCD00236030 CAS-Nummer: 162607-18-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...