2-Acetylphenothiazin, Sigma-Aldrich®

Lieferant: Merck

Synonyme: Methylphenothiazin-2-ylketon

M-Clarity™ Qualitätsstufe = MQ100

175226-100G 175226-25G
SIAL175226-100GEA 689 EUR
SIAL175226-100G SIAL175226-25G
2-Acetylphenothiazin, Sigma-Aldrich®
2-Acetylphenothiazin
Formel: C₁₄H₁₁NOS
Molecular Weight: 241,31 g/mol
MDL: MFCD00005017
CAS-Nummer: 6631-94-3
EINECS: 229-626-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR