tri-Ammoniumhexafluoraluminat 98%

Lieferant: Thermo Scientific
Gefahr

Synonyme: Ammonium hexafluoroaluminate

B20582.09 B20582.18
B20582.09EA 25.9 EUR
B20582.09 B20582.18
tri-Ammoniumhexafluoraluminat 98%
tri-Ammoniumhexafluoraluminat

Purity: 98% CAS No.: 7784-19-2 Molecular Formula: (NH4)3AlF6 Formula Weight: 195.09 Odour: Odourless Merck Reference: 14,524 MDL No.: MFCD00042144

Formel: H₁₂AlF₆N₃
Molecular Weight: 195,09 g/mol
Dichte: 1,78 g/cm³ (20 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00042144
CAS-Nummer: 7784-19-2
EINECS: 232-052-7
UN: 3288
ADR: 6.1,III
Merck Index: 14,524

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR