2,3,4,5,6-Pentafluorstyrol

Lieferant: FLUOROCHEM

006991-5G 006991-25G 006991-10G 006991-50G
FLUO006991-5GEA 45.9 EUR
FLUO006991-5G FLUO006991-25G FLUO006991-10G FLUO006991-50G
2,3,4,5,6-Pentafluorstyrol
2,3,4,5,6-Pentafluorstyrol
Formel: C₈H₃F₅
Molecular Weight: 194,1 g/mol
Siedepunkt: 62 °C (1013 hPa)
Dichte: 1,4295 g/cm³ (20 °C)
MDL: MFCD00000300
CAS-Nummer: 653-34-9
EINECS: 211-500-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR