Phenolphthalein 2% in Ethanol, vergällt, 90%

Lieferant: Bernd Kraft
Gefahr

Synonyme: 3,3-Bis(4-hydroxyphenyl)phthalid, 3,3-Bis(4-hydroxyphenyl)-1(3H)-isobenzofuranon

03956.3700 03956.1700 03956.2700 03956.4700
KRAF03956.3700EA 55.4 EUR
KRAF03956.3700 KRAF03956.1700 KRAF03956.2700 KRAF03956.4700
Phenolphthalein 2% in Ethanol, vergällt, 90%
Phenolphthalein
Formel: C₂₀H₁₄O₄
Molecular Weight: 318.33 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00005913
CAS-Nummer: 77-09-8
EINECS: 201-004-7
UN: 1170
ADR: 3 II
Merck Index: 14,07243
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8d81b539a32ba7c40a473e6afbb8e0dae525d637&vid=91bf141a7ea82baf5df563f73967884d2a4b5d79&prid=web&clid=SAPDAM

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR