(±)-trans-1,2-Diaminocyclohexan 99%
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
Purity: 99%
CAS No.: 1121-22-8
Molecular Formula: C6H14N2
Formula Weight: 114.19
Storage Temperature: 2°C to 8°C
Flash Point: 68°(154°F)
Refractive Index: 1.4890
Odour: Amine-like
Melting Point: 14-15°
Boiling Point: 89-90°/14mm
Beilstein Reference: 3193807
UN No.: 2735
MDL No.: MFCD00063747
Formel:
C₆H₁₄N₂ Siedepunkt: 89…90 °C (14 mmHg) Schmelzpunkt: 14…15 °C Dichte: 0,951 Flammpunkt: 68 °C (154 °F) |
MDL:
MFCD00063747 CAS-Nummer: 1121-22-8 ADR: 8,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...