Anti-BMS-734016 IMC

Lieferant: ProSci
PRSI10-199EA 627 EUR
PRSI10-199
Anti-BMS-734016 IMC
Antikörper
Type: Primary
Antigen:
Clonality: Recombinant
Clone:
Conjugation: Unconjugated
Epitope:
Host:
Isotype:
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR