Anti-CD3e Mouse Monoclonal Antibody [clone: SK7] (APC (Allophycocyanin)/iFluor® 750)
Lieferant: AAT Bioquest
AATB100331G1EA 887 EUR
AATB100331G1 AATB100331G2 AATB100331G0
Anti-CD3e Mouse Monoclonal Antibody [clone: SK7] (APC (Allophycocyanin)/iFluor® 750)
Antikörper
The SK7 monoclonal antibody reacts with human CD3e, a 20 kD member of the Ig superfamily often found on the surface of t cells, tregs and nkt cells.
  • Particularly useful for flow cytometry
This antibody was purified through affinity chromatography.
Bitte beachten For research use only.
Type: Primary
Antigen: CD3e
Clonality: Monoclonal
Clone: SK7
Conjugation: APC (Allophycocyanin)/iFluor® 750
Epitope:
Host: Mouse
Isotype:
Reactivity: Human