Anti-CD5 Mouse Monoclonal Antibody [clone: L17F12] (iFluor® 700)

Lieferant: AAT Bioquest
AATB100510J1EA 1390 EUR
AATB100510J1 AATB100510J0
Anti-CD5 Mouse Monoclonal Antibody [clone: L17F12] (iFluor® 700)
Antikörper
L17F12 is an anti-human monoclonal antibody that is specific for the CD5 antigen.

  • Particularly useful for flow cytometry

This antibody was purified through affinity chromatography.

Bitte beachten For research use only.

Type: Primary
Antigen: CD5
Clonality: Monoclonal
Clone: L17F12
Conjugation: iFluor® 700
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR