Sodium-3-mercapto-1-propanesulphonate 90%

Lieferant: Apollo Scientific

Synonyme: Sodium 3-mercaptopropanesulphonate, 3-Mercapto-1-propanesulphonic acid Sodium Salt

OR1011021-2.5K OR1011021-500G
APOSOR1011021-2.5KEA 870 EUR
APOSOR1011021-2.5K APOSOR1011021-500G
Sodium-3-mercapto-1-propanesulphonate 90%
Sodium-3-mercapto-1-propanesulphonate
Formel: C₃H₈O₃S₂
Molecular Weight: 178.20 g/mol
Schmelzpunkt: 220 °C
MDL: MFCD00043148
CAS-Nummer: 17636-10-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR