Neocuproin Hemihydrat ≥98%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
2,9-Dimethyl-1,10-phenanthrolin Hemihydrat
A11398.06
A11398.14
A11398.22
A11398.06EA
58.7
EUR
A11398.06
A11398.14
A11398.22
Neocuproin Hemihydrat ≥98%
Neocuproin Hemihydrat
Purity: 98+%
CAS No.: 34302-69-7
Molecular Formula: C14H12N2.0.5H2O
Formula Weight: 217.27 (208.26anhy)
Melting Point: 160-164°
Beilstein Reference: 153179
Merck Reference: 14,6449
MDL No.: MFCD00149306
Formel:
C₂₈H₂₆N₄O Molecular Weight: 434,53 g/mol Schmelzpunkt: 160…164 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00149306 CAS-Nummer: 34302-69-7 EINECS: 207-601-9 Merck Index: 14,6449 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...