Neocuproine hydrochloride hydrate, kristallin, Bioreagenzien Reagens für Kupfer, Sigma-Aldrich®
Lieferant: Merck
|
Achtung
|
Synonyme:
2,9-Dimethyl-1,10-phenanthroline hydrochloride hydrate
M-Clarity™ Qualitätsstufe = MQ200
N1626-25G
N1626-5G
SIALN1626-25GEA
481
EUR
SIALN1626-25G
SIALN1626-5G
Neocuproine hydrochloride hydrate, kristallin, Bioreagenzien Reagens für Kupfer, Sigma-Aldrich®
Neocuproine hydrochloride hydrate
Suitable for determination of copper.
Formel:
C₁₄H₁₂N₂·HCl·xH₂O Molecular Weight: 244.72 g/mol (anhy) Schmelzpunkt: (Zers., Lit.) |
MDL:
MFCD00150062 CAS-Nummer: 332360-00-6 EINECS: 230-732-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...