Methyl-5-nitrosalicylat ≥98%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
Methyl 2-hydroxy-5-nitrobenzoate
B25278.09
B25278.18
B25278.30
B25278.09EA
36.6
EUR
B25278.09
B25278.18
B25278.30
Methyl-5-nitrosalicylat ≥98%
Methyl-5-nitrosalicylat
Purity: 98%
CAS No.: 17302-46-4
Molecular Formula: C8H7NO5
Formula Weight: 197.15
Melting Point: 114-117°
MDL No.: MFCD00024695
Formel:
C₈H₇NO₅ Molecular Weight: 197.15 g/mol Schmelzpunkt: 114…117 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00024695 CAS-Nummer: 17302-46-4 EINECS: 241-330-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...