4,7-Dimethyl-1,10-phenanthrolin ≥98%
Lieferant: Thermo Scientific
|
Achtung
|
A17779.01
A17779.03
A17779.06
A17779.01EA
34.2
EUR
A17779.01
A17779.03
A17779.06
4,7-Dimethyl-1,10-phenanthrolin ≥98%
4,7-Dimethyl-1,10-phenanthroline
Purity: 98%
CAS No.: 3248-05-3
Molecular Formula: C14H12N2
Formula Weight: 208.26
Melting Point: 193-195°
MDL No.: MFCD00004979
Formel:
C₁₄H₁₂N₂ Schmelzpunkt: 193…195 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00004979 CAS-Nummer: 3248-05-3 EINECS: 221-827-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...