Methyl-4-chlor-3-nitrobenzoat

Lieferant: Thermo Scientific

Synonyme: 4-Chlor-3-nitrobenzoesäuremethylester

BTB06787.250 BTB06787.1
MAYBBTB06787.250EA 0 EUR
MAYBBTB06787.250 MAYBBTB06787.1
Methyl-4-chlor-3-nitrobenzoat
Methyl-4-chlor-3-nitrobenzoat
Formel: C₈H₆ClNO₄
Molecular Weight: 215,59 g/mol
MDL: MFCD00016989
CAS-Nummer: 14719-83-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR