Hexaammincobalt(III)chlorid 99%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
Cobalt(III)hexamminchlorid
A15470.14
A15470.22
A15470.36
A15470.14EA
47.9
EUR
A15470.14
A15470.22
A15470.36
Hexaammincobalt(III)chlorid 99%
Hexaamminecobalt trichloride
Purity: 99%
CAS No.: 10534-89-1
Molecular Formula: H18Cl3CoN6
Formula Weight: 267.48
Odour: Odourless
Merck Reference: 14,4675
MDL No.: MFCD00036304
Formel:
H₁₈CoCl₃N₆ Molecular Weight: 267,47 g/mol Dichte: 1,71 Lagertemperatur: Raumtemperatur |
MDL:
MFCD00036304 CAS-Nummer: 10534-89-1 EINECS: 234-103-9 Merck Index: 14,4675 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...