Hexaammincobalt(III)chlorid, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
Cobalt(III)hexamminchlorid
M-Clarity™ Qualitätsstufe = MQ200
H7891-10G
H7891-25G
H7891-5G
SIALH7891-10GEA
181
EUR
SIALH7891-10G
SIALH7891-25G
SIALH7891-5G
Hexaammincobalt(III)chlorid, Sigma-Aldrich®
Hexaamminecobalt trichloride
Formel:
H₁₈CoCl₃N₆ Molecular Weight: 267,47 g/mol Dichte: 1,702 g/cm³ (20 °C) |
MDL:
MFCD00036304 CAS-Nummer: 10534-89-1 EINECS: 234-103-9 Merck Index: 12,04711 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...