Polymyxin B-Sulfat

Lieferant: PanReac AppliChem

A0890.0001 A0890.0010
A0890.0001EA 50.5 EUR
A0890.0001 A0890.0010
Polymyxin B-Sulfat
Polymyxin B sulphate
Formel: C₅₅H₉₆N₁₆O₁₃·2H₂SO₄
Molecular Weight: 1385,61 g/mol
MDL: MFCD00131991
CAS-Nummer: 1405-20-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR