ReadiUse™ Annexin Binding Buffer, Optimised for Imaging and Flow Cytometry
Lieferant: AAT BIOQUEST
22870
22871
AATB22870EA
111
EUR
AATB22870
AATB22871
ReadiUse™ Annexin Binding Buffer, Optimised for Imaging and Flow Cytometry
Puffer
Assay-Puffer
The ReadiUse™ Annexin Binding Buffer *Optimised for Imaging and Flow Cytometry* is an isotonic buffer designed to promote the labeling of apoptotic cells with annexin V conjugates.
- Provided as a 10X concentrate
- Saves time
- pH of 7.4
Prior to annexin V staining, we ecommend diluting the 10X concentrate to a 1X working solution using distilled water.
Bitte beachten For research use only.