Anti-Iba1 Rabbit Polyclonal Antibody

Lieferant: ANTIBODIES.COM
A104332-100 A104332-50
ANTIA104332-100EA 520 EUR
ANTIA104332-100 ANTIA104332-50
Anti-Iba1 Rabbit Polyclonal Antibody
Antikörper
Rabbit polyclonal antibody to Iba1 for WB, ICC/IF and IHC with samples derived from human, rat and mouse.

  • Validated Applications: WB, ICC/IF, IHC
  • Recommended Dilutions: WB: 1:1000 to 1:5000, ICC/IF: 1:2000 to 1:5000

Type: Primary
Antigen: Iba1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Rat;Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR