iFluor® 830 succinimidyl ester
Lieferant: AAT BIOQUEST
1384
71400
71580
71530
AATB1384EA
464
EUR
AATB1384
AATB71400
AATB71580
AATB71530
iFluor® 830 succinimidyl ester
iFluor® 830 succinimidyl ester
iFluor® 830 dye emission is a unique color for spectrum flow cytometry as it is well separated from commonly used far-red fluorophores such as Cy5, Cy7, or allophycocyanin (APC), facilitating multicolor analysis.
- Stable for 12 months
In vivo fluorescence imaging uses a sensitive camera to detect the fluorescence emission from fluorophores in whole-body living small animals.
Bitte beachten For Research Use Only