Propidiumiodid 1 mg/ml aqueous solution
Lieferant: AAT BIOQUEST
Synonyme: 3,8-Diamino-5-(3-(diethylmethylammonio)propyl)-6-phenylphenanthridiniumdiiodid
17586 17585
AATB17586EA 135 EUR
AATB17586 AATB17585
Propidiumiodid 1 mg/ml aqueous solution
Propidiumiodid
Propidium iodide (PI) is a red-fluorescent dye widely used for staining and identifying dead cell populations in flow cytometry and fluorescence microscopy.
  • Stable for 6 months
PCR (polymerase chain reaction) is a laboratory technique used to amplify DNA (deoxyribonucleic acid) segments.
Bitte beachten For Research Use Only
Formel: C₂₇H₃₄I₂N₄
Molecular Weight: 668.4 g/mol
MDL: MFCD00011921
CAS-Nummer: 25535-16-4