(4-Nitrophenyl)-β-D-glucopyranosid 99%
Lieferant: Thermo Scientific
Synonyme:
(4-Nitrophenyl)-β-D-glucosid
303110010
303110050
303110250
ACRO303110010EA
89.1
EUR
ACRO303110010
ACRO303110050
ACRO303110250
(4-Nitrophenyl)-β-D-glucopyranosid 99%
(4-Nitrophenyl)-β-D-glucopyranosid
Formel:
C₁₂H₁₅NO₈ Molecular Weight: 301,25 g/mol Schmelzpunkt: 165…168 °C Lagertemperatur: Gefrierschrank |
MDL:
MFCD00006593 CAS-Nummer: 2492-87-7 EINECS: 219-661-3 |
Spezifikation Testergebnisse
Appearance | White to yellow Powder |
Infrared spectrum | Conforms |
HPLC | ≥98.5 % (ELSD) |
Specific optical rotation | -96° to -117° (20°C, 589 nm) (c=1, water) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...