(4-Nitrophenyl)-β-D-glucopyranosid ≥98%
Lieferant: Thermo Scientific
Synonyme:
(4-Nitrophenyl)-β-D-glucosid
L10154.03
L10154.06
L10154.03EA
69.7
EUR
L10154.03
L10154.06
(4-Nitrophenyl)-β-D-glucopyranosid ≥98%
(4-Nitrophenyl)-β-D-glucopyranosid
Purity: 98+%
CAS No.: 2492-87-7
Molecular Formula: C12H15NO8
Formula Weight: 301.25
Storage Temperature: 2°C to 8°C
Melting Point: 166-167°
Beilstein Reference: 92211
Optical Rotation: -103° (c=1 in water)
MDL No.: MFCD00006593
Formel:
C₁₂H₁₅NO₈ Molecular Weight: 301,25 g/mol Schmelzpunkt: 166…167 °C Lagertemperatur: Gefrierschrank |
MDL:
MFCD00006593 CAS-Nummer: 2492-87-7 EINECS: 219-661-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...