N-(3-Dimethylaminopropyl)-N'-ethylcarbodiimid Hydrochlorid 98+%
Lieferant: Thermo Scientific
Gefahr
Synonyme: 1-(3-Dimethylaminopropyl)-3-ethylcarbodiimid Hydrochlorid, EDAC HCl, WSC HCl, EDAC Hydrochlorid, N-(3-Dimethylaminopropyl)-N'-ethylcarbodiimid Monohydrochlorid, N'-(Ethylkohlenstoffimidoyl)-N,N-dimethylpropan-1,3-diaminmonohydrochlorid, EDC Hydrochlorid, EDC HCl
A10807.14 A10807.06 A10807.03
A10807.14EA 308 EUR
A10807.14 A10807.06 A10807.03
N-(3-Dimethylaminopropyl)-N'-ethylcarbodiimid Hydrochlorid 98+%
N-(3-Dimethylaminopropyl)-N'-ethylcarbodiimid Hydrochlorid
Purity: 98+% CAS No.: 25952-53-8 Molecular Formula: C8H17N3.HCl Formula Weight: 191.70 Storage Temperature: 2°C to 8°C Physical Form: Powder Sensitivity: Moisture Sensitive Appearance: White Melting Point: ca 113° Beilstein Reference: 5764110 MDL No.: MFCD00012503
Formel: C₈H₁₇N₃·HCl
Molecular Weight: 191,7 g/mol
Schmelzpunkt: ∼ 113 °C
Lagertemperatur: Kühlschrank
MDL: MFCD00012503
CAS-Nummer: 25952-53-8
EINECS: 247-361-2
UN: 2811
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=25d8bf2864bcf21ffe6dcca6f22388d1fe427e22&vid=9080dd6b66a1622ef32a2da2138f85e44cc881b8&prid=web&clid=SAPDAM