4-(Methylsulfonyl)benzylbromid tech.
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
Synonyme:
α-Brom-4-(methylsulfonyl)toluol
357580025
357580100
ACRO357580025EA
90.8
EUR
ACRO357580025
ACRO357580100
4-(Methylsulfonyl)benzylbromid tech.
4-(Methylsulfonyl)benzylbromid
Formel:
C₈H₉BrO₂S Molecular Weight: 249.13 Schmelzpunkt: 82…86 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00185849 CAS-Nummer: 53606-06-7 UN: 3261 ADR: 8 III |
Spezifikation Testergebnisse
Appearance | Beige to brown Crystalline powder |
Infrared spectrum | Conforms |
GC | ≥60 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...