N,N-Dimethyl-1,3-phenylendiammoniumdichlorid, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
N1,N1-Dimethylbenzene-1,3-diamine dihydrochloride, N,N-Dimethyl-1,3-phenylenediamine dihydrochloride, N,N-Dimethyl-m-phenylenediamine dihydrochloride
M-Clarity™ Qualitätsstufe = MQ100
219223-25G
219223-5G
SIAL219223-25GEA
206
EUR
SIAL219223-25G
SIAL219223-5G
N,N-Dimethyl-1,3-phenylendiammoniumdichlorid, Sigma-Aldrich®
N,N-Dimethyl-1,3-phenylendiammoniumdichlorid
Formel:
C₈H₁₄Cl₂N₂ Molecular Weight: 209,12 g/mol Schmelzpunkt: 218 °C |
MDL:
MFCD00012971 CAS-Nummer: 3575-32-4 EINECS: 222-693-0 UN: 2811 ADR: 6.1,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...