N,N-Dimethyl-1,4-phenylendiammoniumsulfat 98%
Lieferant: Thermo Scientific
|
Gefahr
|
Synonyme:
N,N-Dimethylbenzol-1,4-diammoniumsulfat, N,N-Dimethyl-p-phenylendiammoniumsulfat
A14175.14
A14175.22
A14175.36
A14175.14EA
66.4
EUR
A14175.14
A14175.22
A14175.36
N,N-Dimethyl-1,4-phenylendiammoniumsulfat 98%
N,N-Dimethyl-1,4-phenylendiammoniumsulfat
Purity: 98%
CAS No.: 536-47-0
Molecular Formula: C8H12N2.H2SO4
Formula Weight: 234.28
Odour: Odourless
Melting Point: 200-205° dec.
Beilstein Reference: 4612278
UN No.: 2811
MDL No.: MFCD00012992
Formel:
C₈H₁₄N₂O₄S Molecular Weight: 234,28 g/mol Schmelzpunkt: 200…205 °C (Zersetzung) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00012992 CAS-Nummer: 536-47-0 EINECS: 208-636-2 UN: 2811 ADR: 6.1,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...