N,N-Dimethyl-1,4-phenylendiammoniumsulfat, Sigma-Aldrich®
Lieferant: Merck
|
Gefahr
|
Synonyme:
N,N-Dimethylbenzol-1,4-diammoniumsulfat, N,N-Dimethyl-p-phenylendiammoniumsulfat
M-Clarity™ Qualitätsstufe = MQ200
186384-100G
186384-25G
SIAL186384-100GEA
204
EUR
SIAL186384-100G
SIAL186384-25G
N,N-Dimethyl-1,4-phenylendiammoniumsulfat, Sigma-Aldrich®
N,N-Dimethyl-1,4-phenylendiammoniumsulfat
Formel:
C₈H₁₄N₂O₄S Molecular Weight: 234,28 g/mol Lagertemperatur: Kühlschrank |
MDL:
MFCD00012992 CAS-Nummer: 536-47-0 EINECS: 208-636-2 UN: 2811 ADR: 6.1,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...