2-Methoxy-1-naphthaldehyd ≥99%

Lieferant: Thermo Fisher Scientific

B20483.06 B20483.14 B20483.22
B20483.06EA 27.8 EUR
B20483.06 B20483.14 B20483.22
2-Methoxy-1-naphthaldehyd ≥99%
2-Methoxy-1-naphthaldehyd

Purity: 99% CAS No.: 5392-12-1 Molecular Formula: C12H10O2 Formula Weight: 186.21 Melting Point: 81-85° Boiling Point: 204-206°/18mm MDL No.: MFCD00004004

Formel: C₁₂H₁₀O₂
Molecular Weight: 186.21 g/mol
Siedepunkt: 204…206 °C (18 mmHg)
Schmelzpunkt: 81…85 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00004004
CAS-Nummer: 5392-12-1
EINECS: 226-392-5

Bestellen

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR