3-[(3-Cholamidopropyl)dimethylammonio]-1-propansulfonat (CHAPS)
Lieferant: Uptima
Synonyme:
CHAPS
UP333514
UP333515
UP33351A
UPTIUP333514EA
0
EUR
UPTIUP333514
UPTIUP333515
UPTIUP33351A
3-[(3-Cholamidopropyl)dimethylammonio]-1-propansulfonat (CHAPS)
3-[(3-Cholamidopropyl)dimethylammonio]-1-propansulfonat (CHAPS)
Formel:
C₃₂H₅₈N₂O₇S Schmelzpunkt: 116 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD00081083 CAS-Nummer: 75621-03-3 Merck Index: 13,02052 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...