3-[(3-Cholamidopropyl)dimethylammonio]-1-propansulfonat (CHAPS), Millipore®
Lieferant: Merck Millipore Calbiochem
Synonyme: CHAPS
220201-1 220201-10 220201-25 220201-5 220201-1KG 220201-250GM
220201-1EA 84.2 EUR
220201-1 220201-10 220201-25 220201-5 220201-1KG 220201-250GM
3-[(3-Cholamidopropyl)dimethylammonio]-1-propansulfonat (CHAPS), Millipore®
3-[(3-Cholamidopropyl)dimethylammonio]-1-propansulfonat (CHAPS)
Formel: C₃₂H₅₈N₂O₇S
Molecular Weight: 614,89 g/mol
Schmelzpunkt: 116 °C
Lagertemperatur: Kühlschrank
MDL: MFCD00081083
CAS-Nummer: 75621-03-3
Merck Index: 13,02052