3-[(3-Cholamidopropyl)dimethylammonio]-1-propansulfonat (CHAPS) Hydrat, Sigma-Aldrich®
Lieferant: Merck
M-Clarity™ Qualitätsstufe = MQ200
226947-1G
226947-5G
SIAL226947-1GEA
66.6
EUR
SIAL226947-1G
SIAL226947-5G
3-[(3-Cholamidopropyl)dimethylammonio]-1-propansulfonat (CHAPS) Hydrat, Sigma-Aldrich®
3-[(3-Cholamidopropyl)dimethylammonio]-1-propansulfonat (CHAPS) Hydrat
Formel:
C₃₂H₅₈N₂O₇S·H₂O Molecular Weight: 632,9 g/mol Lagertemperatur: Kühlschrank |
CAS-Nummer:
313223-04-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...