3-((3-Cholamidopropyl)dimethylammonio)-2-hydroxy-1-propansulfonat

Lieferant: Uptima
Achtung

Synonyme: 3-((3-Cholamidopropyl)dimethylammonio)-2-hydroxypropansulfonat, CHAPSO

UP356392 UP356393
UPTIUP356392EA 0 EUR
UPTIUP356392 UPTIUP356393
3-((3-Cholamidopropyl)dimethylammonio)-2-hydroxy-1-propansulfonat
(3-((3-Cholamidopropyl)dimethylammonio)-2-hydroxy-1-propanesulphonate)
Formel: C₃₂H₅₈N₂O₈S
Schmelzpunkt: 184…186 °C
Lagertemperatur: Kühlschrank
CAS-Nummer: 82473-24-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR