4-Fluor-1-naphthoesäure

Lieferant: FLUOROCHEM

CFF1133.1 CFF1133.10 006290-1G 006290-5G
CHMPCFF1133.1EA 522 EUR
CHMPCFF1133.1 CHMPCFF1133.10 FLUO006290-1G FLUO006290-5G
4-Fluor-1-naphthoesäure
4-Fluor-1-naphthoesäure
Formel: C₁₁H₇FO₂
MDL: MFCD00004011
CAS-Nummer: 573-03-5
EINECS: 209-346-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR