4-Hydroxy-1-naphthaldehyd, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

131067-1G 131067-5G
SIAL131067-1GEA 47.9 EUR
SIAL131067-1G SIAL131067-5G
4-Hydroxy-1-naphthaldehyd, Sigma-Aldrich®
4-Hydroxy-1-naphthaldehyd
Formel: C₁₁H₈O₂
Molecular Weight: 172,18 g/mol
MDL: MFCD00053486
CAS-Nummer: 7770-45-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR