5-Amino-2-naphthalinsulfonsäure, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 5-Amino-β-naphthalinsulfonsäure, 5-Aminonaphthalin-2-sulfonsäure, 1-Naphthylamin-6-sulfonsäure, 2-Naphthylamin-5-sulfonsäure

M-Clarity™ Qualitätsstufe = MQ100

285072-100G 285072-5G
SIAL285072-100GEA 208 EUR
SIAL285072-100G SIAL285072-5G
5-Amino-2-naphthalinsulfonsäure, Sigma-Aldrich®
5-Amino-2-naphthalenesulphonic acid
Formel: C₁₀H₉NO₃S
Molecular Weight: 223.25 g/mol
MDL: MFCD00004030
CAS-Nummer: 119-79-9
EINECS: 204-351-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR