X-Gal (5-Brom-4-chlor-3-indolyl-ß-D-galactopyranosid)
Lieferant: BACHEM BIOCHEMICA
Synonyme:
BCIG
4000924.0250
4000924.1000
BACE4000924.0250EA
0
EUR
BACE4000924.0250
BACE4000924.1000
X-Gal (5-Brom-4-chlor-3-indolyl-ß-D-galactopyranosid)
X-Gal (5-Brom-4-chlor-3-indolyl-ß-D-galactopyranosid)
Formel:
C₁₄H₁₅BrClNO₆ Schmelzpunkt: 237…239 °C Lagertemperatur: Gefrierschrank |
MDL:
MFCD00005666 CAS-Nummer: 7240-90-6 EINECS: 230-640-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...