Decyl-β-D-glucopyranosid, Sigma-Aldrich®

Lieferant: Merck
Achtung

Synonyme: Decyl-β-D-glucosid

M-Clarity™ Qualitätsstufe = MQ200

D5394-1G D5394-5G
SIALD5394-1GEA 394 EUR
SIALD5394-1G SIALD5394-5G
Decyl-β-D-glucopyranosid, Sigma-Aldrich®
Decyl-β-D-glucopyranosid
Formel: C₁₆H₃₂O₆
Molecular Weight: 320,43 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00063297
CAS-Nummer: 58846-77-8
EINECS: 261-469-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR