Bis(trimethylsilyl)carbodiimid
Lieferant: CHEMPUR SILANE
SILS02575.5
SILS02575.25
SILNSILS02575.5EA
0
EUR
SILNSILS02575.5
SILNSILS02575.25
Bis(trimethylsilyl)carbodiimid
Bis(trimethylsilyl)carbodiimid
Formel:
C₇H₁₈N₂Si₂ Molecular Weight: 186,4 g/mol Siedepunkt: 164 °C (1013 hPa) Schmelzpunkt: –23 °C Dichte: 0,8211 g/cm³ (25 °C) |
MDL:
MFCD00051538 CAS-Nummer: 1000-70-0 EINECS: 213-673-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...