Lithiumaluminat

Lieferant: Thermo Fisher Scientific

Synonyme: Aluminiumlithiumdioxid, Aluminiumlithiumoxid, Lithiumaluminiumoxid

13412.18 13412.30 13412.A1
13412.18EA 101 EUR
13412.18 13412.30 13412.A1
Lithiumaluminat
Lithium aluminate

CAS No.: 12003-67-7 Molecular Formula: LiAlO2 Formula Weight: 65.92 Physical Form: Powder Density: 2.62 Odour: Odourless MDL No.: MFCD00016172

Formel: AlLiO₂
Molecular Weight: 65.92 g/mol
Dichte: 2.62
Lagertemperatur: Raumtemperatur
MDL: MFCD00016172
CAS-Nummer: 12003-67-7
EINECS: 234-434-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR