Natriumaluminat, tech.
Lieferant: Thermo Scientific
|
Gefahr
|
Synonyme:
Natriumaluminiumoxid, Aluminiumnatriumdioxid, Aluminiumnatriumoxid
35453.22
35453.36
35453.22EA
57.1
EUR
35453.22
35453.36
Natriumaluminat, tech.
Sodium aluminate
CAS No.: 1302-42-7
Molecular Formula: Al2O3.Na2O
Formula Weight: 163.94
Physical Form: Crystalline
Density: 1.52
Sensitivity: Hygroscopic
Appearance: White
Odour: Odourless
Solubility: Insoluble in alcohol, soluble in water
UN No.: 2812
MDL No.: MFCD00801010
Formel:
NaAlO₂ Molecular Weight: 81,97 g/mol Schmelzpunkt: 1800 °C Dichte: 1,52 Lagertemperatur: Raumtemperatur |
MDL:
MFCD00801010 CAS-Nummer: 1302-42-7 EINECS: 215-100-1 UN: 2812 ADR: 8,III Merck Index: 12,08715 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...