Nonyl-β-D-glucopyranosid, Sigma-Aldrich®

Lieferant: Merck

Synonyme: Nonyl-β-D-glucosidNG

M-Clarity™ Qualitätsstufe = MQ200

SIALN7507-1GEA 265 EUR
SIALN7507-1G SIALN7507-5G
Nonyl-β-D-glucopyranosid, Sigma-Aldrich®
Nonyl-β-D-glucopyranosid
Formel: C₁₅H₃₀O₆
Molecular Weight: 306,4 g/mol
MDL: MFCD00063300
CAS-Nummer: 69984-73-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR