Nonyl-β-D-glucopyranosid

Lieferant: Uptima

Synonyme: Nonyl-β-D-glucosid, NG

UP602071 UP602072
UPTIUP602071EA 0 EUR
UPTIUP602071 UPTIUP602072
Nonyl-β-D-glucopyranosid
Nonyl-β-D-glucopyranosid
Formel: C₁₅H₃₀O₆
Molecular Weight: 306,4 g/mol
MDL: MFCD00063300
CAS-Nummer: 69984-73-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR