Nonyl-β-D-glucopyranosid
Lieferant: BACHEM BIOCHEMICA
Synonyme:
Nonyl-β-D-glucosid, NG
4026563.0001
4026563.0005
BACE4026563.0001EA
0
EUR
BACE4026563.0001
BACE4026563.0005
Nonyl-β-D-glucopyranosid
Nonyl-β-D-glucopyranosid
Formel:
C₁₅H₃₀O₆ Molecular Weight: 306,4 g/mol |
MDL:
MFCD00063300 CAS-Nummer: 69984-73-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...